missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(3-Thienyl)acrylic acid, 98%

Catalog No. p-7027942
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2501706 5 g
AAB2501714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2501706 Supplier Thermo Scientific Chemicals Supplier No. B2501706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 102696-71-9 | C7H6O2S | 154.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102696-71-9
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.183
MDL Number MFCD00014535
InChI Key VYRYYUKILKRGDN-OWOJBTEDSA-N
Synonym 3-3-thienyl acrylic acid, 3-thien-3-ylacrylic acid, 3-thiophen-3-yl acrylic acid, trans-3-3-thienyl acrylic acid, 2e-3-thiophen-3-yl prop-2-enoic acid, e-3-thiophen-3-yl acrylic acid, 3-thiophen-3-yl prop-2-enoic acid, trans-3-3-thienyl acrylic, 2-propenoic acid, 3-3-thienyl-, 2e, 2e-3-3-thienyl prop-2-enoic acid
PubChem CID 735793
IUPAC Name (E)-3-thiophen-3-ylprop-2-enoic acid
SMILES C1=CSC=C1C=CC(=O)O

Specifications

Melting Point 150°C to 154°C
Quantity 5 g
Beilstein 112245
Formula Weight 154.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material trans-3-(3-Thienyl)acrylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-800-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only