missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Tetradecenoyl-L-carnitine, Analytical Standard, ≥95.0% (LC/MS), MilliporeSigma™ Supelco™
Specifications
Specifications
| Linear Formula | C21H39NO4 |
| Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-[[(2 E)-1-oxo-2-tetradecen-1-yl]oxy]-1-propanaminium inner salt; trans-2-Myristenoyl-L-carnitine; C14:1-Carnitine |
| Quantity | 10 mg |
| Formula Weight | 369.54 |
| Percent Purity | ≥95.0% (LC/MS) |
| Grade | Analytical Standard |