missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Phenylcyclopropylamine hydrochloride, 97%

Catalog No. p-3779417
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130472500 250 mg Glass bottle
AC130470010 1 g Glass bottle
AC130470050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC130472500 Supplier Thermo Scientific Chemicals Supplier No. 130472500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1986-47-6 | C9H11N·HCl | 169.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1986-47-6
Molecular Formula C9H11N·HCl
Molecular Weight (g/mol) 169.66
MDL Number MFCD00063602
InChI Key ZPEFMSTTZXJOTM-VTLYIQCISA-N
Synonym tranylcypromine hydrochloride, trans-2-phenylcyclopropanamine hydrochloride, trans-2-phenylcyclopropylamine hydrochloride, 1r,2s-2-phenyl-cyclopropylamine hydrochloride, 1r,2s-2-phenylcyclopropan-1-amine hydrochloride, tranylcypromine hcl, 1r,2s-2-phenylcyclopropanamine hydrochloride, trans-2-phenyl-cyclopropylamine hydrochloride, dsstox_cid_27761, dsstox_rid_82538
PubChem CID 12345947
IUPAC Name (1R,2R)-2-phenylcyclopropan-1-amine;hydrochloride
SMILES C1C(C1N)C2=CC=CC=C2.Cl

Specifications

Melting Point 162°C to 169°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H5C3H4NH2·HCl
Quantity 250 mg
Beilstein 12, III, 2797
Merck Index 14, 9573
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ethyl acetate,diethyl ether
Formula Weight 169.66
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material trans-2-Phenylcyclopropylamine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only