missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Phenyl-1-cyclohexanol 98.0+%, TCI America™
Supplier: TCI America P11741G
Specifications
| trans-2-Phenyl-1-cyclohexanol | |
| 58°C | |
| 122°C | |
| MFCD00003865 | |
| AAIBYZBZXNWTPP-NWDGAFQWSA-N | |
| (1R,2S)-2-phenylcyclohexan-1-ol | |
| 102793 | |
| ≥98.0% (GC) |
| 2362-61-0 | |
| White | |
| C12H16O | |
| 1 g | |
| C1CCC(C(C1)C2=CC=CC=C2)O | |
| 176.259 | |
| 176.26 | |
| Crystalline Powder |
Chemical Identifiers
| 2362-61-0 | |
| 176.259 | |
| AAIBYZBZXNWTPP-NWDGAFQWSA-N | |
| (1R,2S)-2-phenylcyclohexan-1-ol |
| C12H16O | |
| MFCD00003865 | |
| 102793 | |
| C1CCC(C(C1)C2=CC=CC=C2)O |
Safety and Handling
TSCA : No