missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Phenyl-1-cyclohexanol 98.0+%, TCI America™
$152.86 - $456.70
Chemical Identifiers
| CAS | 2362-61-0 |
|---|---|
| Molecular Formula | C12H16O |
| Molecular Weight (g/mol) | 176.259 |
| MDL Number | MFCD00003865 |
| InChI Key | AAIBYZBZXNWTPP-NWDGAFQWSA-N |
| PubChem CID | 102793 |
| IUPAC Name | (1R,2S)-2-phenylcyclohexan-1-ol |
| SMILES | C1CCC(C(C1)C2=CC=CC=C2)O |
Chemical Identifiers
| 2362-61-0 | |
| 176.259 | |
| AAIBYZBZXNWTPP-NWDGAFQWSA-N | |
| (1R,2S)-2-phenylcyclohexan-1-ol |
| C12H16O | |
| MFCD00003865 | |
| 102793 | |
| C1CCC(C(C1)C2=CC=CC=C2)O |
Specifications
| 2362-61-0 | |
| White | |
| C12H16O | |
| 1 g | |
| C1CCC(C(C1)C2=CC=CC=C2)O | |
| 176.259 | |
| 176.26 | |
| Crystalline Powder |
| 58°C | |
| 122°C | |
| MFCD00003865 | |
| AAIBYZBZXNWTPP-NWDGAFQWSA-N | |
| (1R,2S)-2-phenylcyclohexan-1-ol | |
| 102793 | |
| ≥98.0% (GC) | |
| trans-2-Phenyl-1-cyclohexanol |
Safety and Handling
TSCA : No