missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentenoic acid, 90+%, remainder other isomers

CAS: 13991-37-2 | C5H8O2 | 100.12 g/mol

$128.12 - $504.88

Chemical Identifiers

CAS 13991-37-2
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00002704
InChI Key YIYBQIKDCADOSF-ONEGZZNKSA-N
Synonym trans-2-pentenoic acid, 2-pentenoic acid, e-pent-2-enoic acid, pent-2-enoic acid, 2e-pent-2-enoic acid, 2-pentenoic acid, 2e, beta-ethyl acrylic acid, c2h5ch=chcooh, pentenoic acid, unii-1rg66883cf
PubChem CID 638122
ChEBI CHEBI:38366
IUPAC Name (E)-pent-2-enoic acid
SMILES CC\C=C\C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1268909
View Documents Promotion Details
N/A 10 g
Each for $128.12
Only null left
 
AAA1268918
View Documents Promotion Details
N/A 50 g
Each for $504.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13991-37-2
100.12
YIYBQIKDCADOSF-ONEGZZNKSA-N
638122
(E)-pent-2-enoic acid
C5H8O2
MFCD00002704
trans-2-pentenoic acid, 2-pentenoic acid, e-pent-2-enoic acid, pent-2-enoic acid, 2e-pent-2-enoic acid, 2-pentenoic acid, 2e, beta-ethyl acrylic acid, c2h5ch=chcooh, pentenoic acid, unii-1rg66883cf
CHEBI:38366
CC\C=C\C(O)=O

Specifications

13991-37-2
0.992
102°C (215°F)
C5H8O2
MFCD00002704
1720312
YIYBQIKDCADOSF-ONEGZZNKSA-N
(E)-pent-2-enoic acid
638122
100.12
remainder other isomers
8°C to 10°C
200°C to 203°C
Pungent
1.4515
UN3265
trans-2-pentenoic acid, 2-pentenoic acid, e-pent-2-enoic acid, pent-2-enoic acid, 2e-pent-2-enoic acid, 2-pentenoic acid, 2e, beta-ethyl acrylic acid, c2h5ch=chcooh, pentenoic acid, unii-1rg66883cf
CC\C=C\C(O)=O
100.12
CHEBI:38366
≥90%
trans-2-Pentenoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 237-791-9

RTECSNumber : SB2750000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only