missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentenal, 96%

Catalog No. AAL0610806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0610806 5 g
AAL0610814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0610806 Supplier Thermo Scientific Chemicals Supplier No. L0610806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1576-87-0 | C5H8O | 84.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1576-87-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00009615
InChI Key DTCCTIQRPGSLPT-ONEGZZNKSA-N
Synonym trans-2-pentenal, 2-pentenal, e-2-pentenal, e-pent-2-enal, 3-ethylacrolein, pent-2-enal, 2-pentenal, e, 2-e-pentenal, 3-ethyl-2-propenal, e-pent-2-en-1-al
PubChem CID 5364752
IUPAC Name (E)-pent-2-enal
SMILES CCC=CC=O

Specifications

Density 0.86
Boiling Point 125°C
Flash Point 22°C (71°F)
Odor Fruit-like
Refractive Index 1.443
Quantity 5 g
UN Number UN1989
Beilstein 1719742
Sensitivity Air Sensitive
Formula Weight 84.12
Percent Purity 96%
Chemical Name or Material trans-2-Pentenal
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 216-414-1
RTECSNumber SB1560000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only