missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octenal 96.0+%, TCI America™

Catalog No. p-7134224
Change view
Click to view available options
Quantity:
10 mL
25 mL
Catalog No. Quantity
O017610ML 10 mL
O017625ML 25 mL
2 options

Catalog No. O017610ML

Supplier: TCI America O017610ML

Only null left

Chemical Identifiers

CAS 2548-87-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00007011
InChI Key LVBXEMGDVWVTGY-VOTSOKGWSA-N
Synonym trans-2-octenal, e-oct-2-enal, e-2-octenal, 2-octenal, trans-2-octen-1-al, e-2-octen-1-al, 2-octen-1-al, 2-octenel, 2-octenal, e, 2-octenal, 2e
PubChem CID 5283324
ChEBI CHEBI:61748
IUPAC Name (E)-oct-2-enal
SMILES CCCCCC=CC=O

Specifications

Color Yellow
Boiling Point 78°C
Quantity 10 mL
Formula Weight 126.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Octenal
RTECSNumber RH2145000
TSCA Yes