missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Octen-1-ol 95.0+%, TCI America™
Supplier: TCI America O0247100ML
| Quantity | 100 mL |
|---|
Chemical Identifiers
| 18409-17-1 | |
| 128.215 | |
| AYQPVPFZWIQERS-VOTSOKGWSA-N | |
| 5318599 | |
| CCCCCC=CCO |
| C8H16O | |
| MFCD00014057 | |
| trans-2-octen-1-ol, e-oct-2-en-1-ol, e-2-octen-1-ol, 2-octenol, 2-octen-1-ol, e, 2-octen-1-ol, 2e-octen-1-ol, unii-k54st7v77u, 2-octen-1-ol, 2e, 2e-oct-2-en-1-ol | |
| (E)-oct-2-en-1-ol |
Specifications
| trans-2-Octen-1-ol | |
| Yellow | |
| C8H16O | |
| MFCD00014057 | |
| AYQPVPFZWIQERS-VOTSOKGWSA-N | |
| (E)-oct-2-en-1-ol | |
| 5318599 | |
| ≥95.0% (GC) |
| 18409-17-1 | |
| 92°C | |
| 100 mL | |
| trans-2-octen-1-ol, e-oct-2-en-1-ol, e-2-octen-1-ol, 2-octenol, 2-octen-1-ol, e, 2-octen-1-ol, 2e-octen-1-ol, unii-k54st7v77u, 2-octen-1-ol, 2e, 2e-oct-2-en-1-ol | |
| CCCCCC=CCO | |
| 128.215 | |
| 128.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2393
TSCA : Yes