Learn More
trans-2-Octen-1-ol, 97%
CAS: 18409-17-1 | C8H16O | 128.215 g/mol
Supplier: Thermo Scientific Chemicals A1596014
| Quantity | 25 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 18409-17-1 | |
| 128.215 | |
| AYQPVPFZWIQERS-VOTSOKGWSA-N | |
| 5318599 | |
| CCCCCC=CCO |
| C8H16O | |
| MFCD00014057 | |
| trans-2-octen-1-ol, e-oct-2-en-1-ol, e-2-octen-1-ol, 2-octenol, 2-octen-1-ol, e, 2-octen-1-ol, 2e-octen-1-ol, unii-k54st7v77u, 2-octen-1-ol, 2e, 2e-oct-2-en-1-ol | |
| (E)-oct-2-en-1-ol |
Specifications
| trans-2-Octen-1-ol | |
| 0.85 | |
| 85°C (185°F) | |
| 1.4465 | |
| MFCD00014057 | |
| trans-2-octen-1-ol, e-oct-2-en-1-ol, e-2-octen-1-ol, 2-octenol, 2-octen-1-ol, e, 2-octen-1-ol, 2e-octen-1-ol, unii-k54st7v77u, 2-octen-1-ol, 2e, 2e-oct-2-en-1-ol | |
| CCCCCC=CCO | |
| 128.215 | |
| 128.22 |
| 18409-17-1 | |
| 88°C (11 mmHg) | |
| C8H16O | |
| 25 g | |
| 1720707 | |
| AYQPVPFZWIQERS-VOTSOKGWSA-N | |
| (E)-oct-2-en-1-ol | |
| 5318599 | |
| 97% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
EINECSNumber : 242-288-2
TSCA : Yes
Recommended Storage : Ambient temperatures