missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Nonenal 95.0+%, TCI America™

Catalog No. p-7133833
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04835ML 5 mL
N048325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04835ML Supplier TCI America Supplier No. N04835ML

Chemical Identifiers

CAS 18829-56-6
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00007012
InChI Key BSAIUMLZVGUGKX-BQYQJAHWSA-N
Synonym trans-2-nonenal, 2-nonenal, e-2-nonenal, e-non-2-enal, 3-hexylacrolein, 2-nonenal, 2e, beta-hexylacrolein, 2-nonenal, e, heptylideneacetaldehyde, alpha-nonenyl aldehyde
PubChem CID 5283335
IUPAC Name (E)-non-2-enal
SMILES CCCCCCC=CC=O

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 mL
Formula Weight 140.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Nonenal
RTECSNumber RA8509050
TSCA Yes