CAS RN 18829-56-6
trans-2-Nonenal, 95%, Thermo Scientific™
CAS: 18829-56-6 Molecular Formula: C9H16O Molecular Weight (g/mol): 140.23 MDL Number: MFCD00007012 InChI Key: BSAIUMLZVGUGKX-BQYQJAHWSA-N Synonym: trans-2-nonenal,2-nonenal,e-2-nonenal,e-non-2-enal,3-hexylacrolein,2-nonenal, 2e,beta-hexylacrolein,2-nonenal, e,heptylideneacetaldehyde,alpha-nonenyl aldehyde PubChem CID: 5283335 IUPAC Name: (E)-non-2-enal SMILES: CCCCCCC=CC=O
trans-2-Nonenal, 97%
CAS: 18829-56-6 Molecular Formula: C9H16O Molecular Weight (g/mol): 140.226 MDL Number: MFCD00007012 InChI Key: BSAIUMLZVGUGKX-BQYQJAHWSA-N Synonym: trans-2-nonenal,2-nonenal,e-2-nonenal,e-non-2-enal,3-hexylacrolein,2-nonenal, 2e,beta-hexylacrolein,2-nonenal, e,heptylideneacetaldehyde,alpha-nonenyl aldehyde PubChem CID: 5283335 IUPAC Name: (E)-non-2-enal SMILES: CCCCCCC=CC=O
trans-2-Nonenal 95.0+%, TCI America™
CAS: 18829-56-6 Molecular Formula: C9H16O Molecular Weight (g/mol): 140.226 MDL Number: MFCD00007012 InChI Key: BSAIUMLZVGUGKX-BQYQJAHWSA-N Synonym: trans-2-nonenal,2-nonenal,e-2-nonenal,e-non-2-enal,3-hexylacrolein,2-nonenal, 2e,beta-hexylacrolein,2-nonenal, e,heptylideneacetaldehyde,alpha-nonenyl aldehyde PubChem CID: 5283335 IUPAC Name: (E)-non-2-enal SMILES: CCCCCCC=CC=O
trans-2-Nonenal (contains trans-2-Nonenal Diethyl Acetal) (ca. 10% in Ethanol, ca. 0.57mol/L), TCI America™
CAS: 18829-56-6 Molecular Formula: C9H16O Molecular Weight (g/mol): 140.226 MDL Number: MFCD00007012 InChI Key: BSAIUMLZVGUGKX-BQYQJAHWSA-N Synonym: trans-2-nonenal,2-nonenal,e-2-nonenal,e-non-2-enal,3-hexylacrolein,2-nonenal, 2e,beta-hexylacrolein,2-nonenal, e,heptylideneacetaldehyde,alpha-nonenyl aldehyde PubChem CID: 5283335 IUPAC Name: (E)-non-2-enal SMILES: CCCCCCC=CC=O