missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Nonenal (contains trans-2-Nonenal Diethyl Acetal) (ca. 10% in Ethanol, ca. 0.57mol/L), TCI America™

Catalog No. N043025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N043025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N043025ML Supplier TCI America Supplier No. N043025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18829-56-6
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00007012
InChI Key BSAIUMLZVGUGKX-BQYQJAHWSA-N
Synonym trans-2-nonenal, 2-nonenal, e-2-nonenal, e-non-2-enal, 3-hexylacrolein, 2-nonenal, 2e, beta-hexylacrolein, 2-nonenal, e, heptylideneacetaldehyde, alpha-nonenyl aldehyde
PubChem CID 5283335
IUPAC Name (E)-non-2-enal
SMILES CCCCCCC=CC=O

Specifications

Color Yellow
Quantity 25 mL
UN Number 1170
Formula Weight 140.23
Physical Form Liquid
Chemical Name or Material trans-2-Nonenal (contains trans-2-Nonenal Diethyl Acetal) (ca. 10% in Ethanol, ca. 0.57mol/L)
RTECSNumber RA8509050
TSCA Yes