missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Nonenal, 97%

Catalog No. AAA1810806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1810806 5 g
AAA1810814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1810806 Supplier Thermo Scientific Chemicals Supplier No. A1810806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18829-56-6 | C9H16O | 140.226 g/mol

trans-2-Nonenal reacts with acetic acid hydrazide to produce acetic acid non-2-enylidene-hydrazide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-2-Nonenal reacts with acetic acid hydrazide to produce acetic acid non-2-enylidene-hydrazide.

Solubility
Soluble in Alcohol , and oils. Insoluble in water.

Notes
Air Sensitive. Incompatible with air, oxidizing agents and heat. Protect from heat.

Chemical Identifiers

CAS 18829-56-6
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00007012
InChI Key BSAIUMLZVGUGKX-BQYQJAHWSA-N
Synonym trans-2-nonenal, 2-nonenal, e-2-nonenal, e-non-2-enal, 3-hexylacrolein, 2-nonenal, 2e, beta-hexylacrolein, 2-nonenal, e, heptylideneacetaldehyde, alpha-nonenyl aldehyde
PubChem CID 5283335
IUPAC Name (E)-non-2-enal
SMILES CCCCCCC=CC=O

Specifications

Density 0.846
Boiling Point 98°C to 100°C (12 mmHg)
Flash Point 84°C (184°F)
Refractive Index 1.4403
Quantity 5 g
Sensitivity Air sensitive
Merck Index 14,6676
Solubility Information Soluble in Alcohol ,and oils. Insoluble in water.
Formula Weight 140.22
Percent Purity 97%
Chemical Name or Material trans-2-Nonenal
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 242-609-6
RTECSNumber RA8509050
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only