missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Methyl-3-phenyl-2-propen-1-ol, 95%

Catalog No. AC427050050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427050050 5 g Glass bottle
AC427050250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427050050 Supplier Thermo Scientific Chemicals Supplier No. 427050050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1504-55-8 | C10H12O | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1504-55-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
InChI Key LLNAMUJRIZIXHF-VQHVLOKHSA-N
Synonym 2-methyl-3-phenyl-2-propen-1-ol, beta-methylcinnamyl alcohol, 2-propen-1-ol, 2-methyl-3-phenyl, alpha-methylcinnamyl alcohol, methyl cinnamic alcohol, alpha-methylcinnamic alcohol, alpha methyl cinnamic alcohol, 3-phenyl-2-methyl-propen-2-ol-1, e-2-methyl-3-phenyl-2-propen-1-ol, 2e-2-methyl-3-phenylprop-2-en-1-ol
PubChem CID 6393846
IUPAC Name (E)-2-methyl-3-phenylprop-2-en-1-ol
SMILES CC(=CC1=CC=CC=C1)CO

Specifications

Density 1.0300g/mL
Boiling Point 77°C (0.1 mmHg)
Flash Point 113°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=C(CH3)CH2OH
Quantity 5 g
Specific Gravity 1.03
Formula Weight 148.2
Percent Purity 95%
Chemical Name or Material trans-2-Methyl-3-phenyl-2-propen-1-ol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-128-7

RUO – Research Use Only