accessibility menu, dialog, popup

UserName

CAS RN 1504-55-8

IUPAC Name: (3E)-4-phenylbut-3-en-2-ol
Synonyms: methylcinnamyl alcohol
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: ZIJWGEHOVHJHKB-BQYQJAHWNA-N
SMILES: CC(O)\C=C\C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)