missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Methoxycinnamic Acid 99.0+%, TCI America™

Catalog No. m044925g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M044925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M044925G Supplier TCI America Supplier No. M044925G
Only null left

Chemical Identifiers

CAS 1011-54-7
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00064238
InChI Key FEGVSPGUHMGGBO-VOTSOKGWSA-N
Synonym 2-methoxycinnamic acid, trans-2-methoxycinnamic acid, 3-2-methoxyphenyl acrylic acid, 2e-3-2-methoxyphenyl acrylic acid, 2e-3-2-methoxyphenyl prop-2-enoic acid, o-methoxycinnamic acid, e-3-2-methoxyphenyl acrylic acid, e-3-2-methoxyphenyl-2-propenoic acid, e-o-methoxycinnamic acid, trans-o-methyl-o-coumaric acid
PubChem CID 734154
IUPAC Name (E)-3-(2-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=CC=C1C=CC(=O)O

Specifications

Melting Point 186°C
Quantity 25 g
Formula Weight 178.19
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans-2-Methoxycinnamic Acid