missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Methoxycinnamic acid, 98+%

Catalog No. p-7022410
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1517306 5 g
AAA1517314 25 g
AAA1517322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1517306 Supplier Thermo Scientific Chemicals Supplier No. A1517306
Only null left

CAS: 1011-54-7 | C10H10O3 | 178.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1011-54-7
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00064238
InChI Key FEGVSPGUHMGGBO-VOTSOKGWSA-N
Synonym 2-methoxycinnamic acid, trans-2-methoxycinnamic acid, 3-2-methoxyphenyl acrylic acid, 2e-3-2-methoxyphenyl acrylic acid, 2e-3-2-methoxyphenyl prop-2-enoic acid, o-methoxycinnamic acid, e-3-2-methoxyphenyl acrylic acid, e-3-2-methoxyphenyl-2-propenoic acid, e-o-methoxycinnamic acid, trans-o-methyl-o-coumaric acid
PubChem CID 734154
IUPAC Name (E)-3-(2-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=CC=C1C=CC(=O)O

Specifications

Melting Point 183°C to 186°C
Quantity 5 g
Beilstein 2209712
Formula Weight 178.19
Percent Purity ≥98%
Chemical Name or Material trans-2-Methoxycinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-785-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only