missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99+%

CAS: 28250-45-5 | C7H16NO | 130.21 g/mol

$935.77 - $935.77

Chemical Identifiers

CAS 28250-45-5
Molecular Formula C7H16NO
Molecular Weight (g/mol) 130.21
MDL Number MFCD00145424
InChI Key GCWPGEWXYDEQAY-NKWVEPMBSA-O
Synonym trans-2-hydroxymethyl-1-cyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexyl methanol hydrochloride, trans-2-aminocyclohexyl methanol hydrochloride, trans-2-amino-cyclohexyl-methanol hydrochloride, trans-2-aminocyclohexylmethanol hydrochloride, 1r,2r-2-aminocyclohexyl methan-1-ol, chloride, trans-+/--2-aminocyclohexyl methanol hydrochloride, trans-+/--2-aminocyclohexyl methanol hydrochloride salt, cyclohexanemethanol, 2-amino-, hydrochloride, 1r,2r-rel
PubChem CID 2724654
IUPAC Name [(1R,2R)-2-aminocyclohexyl]methanol;hydrochloride
SMILES [NH3+][C@@H]1CCCC[C@H]1CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC265870010
SDS View Documents
Thermo Scientific Chemicals
265870010
1 g Glass bottle
Each for $935.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

28250-45-5
130.21
GCWPGEWXYDEQAY-NKWVEPMBSA-O
2724654
[NH3+][C@@H]1CCCC[C@H]1CO
C7H16NO
MFCD00145424
trans-2-hydroxymethyl-1-cyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexyl methanol hydrochloride, trans-2-aminocyclohexyl methanol hydrochloride, trans-2-amino-cyclohexyl-methanol hydrochloride, trans-2-aminocyclohexylmethanol hydrochloride, 1r,2r-2-aminocyclohexyl methan-1-ol, chloride, trans-+/--2-aminocyclohexyl methanol hydrochloride, trans-+/--2-aminocyclohexyl methanol hydrochloride salt, cyclohexanemethanol, 2-amino-, hydrochloride, 1r,2r-rel
[(1R,2R)-2-aminocyclohexyl]methanol;hydrochloride

Specifications

28250-45-5
White
99% min. (Argentometry)
C7H16NO
1 g
Solubility in water: soluble. Other solubilities: soluble in methanol
[NH3+][C@@H]1CCCC[C@H]1CO
130.21
165.66
Crystalline Powder
150°C to 155°C
Authentic
Glass bottle
MFCD00145424
trans-2-hydroxymethyl-1-cyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexyl methanol hydrochloride, trans-2-aminocyclohexyl methanol hydrochloride, trans-2-amino-cyclohexyl-methanol hydrochloride, trans-2-aminocyclohexylmethanol hydrochloride, 1r,2r-2-aminocyclohexyl methan-1-ol, chloride, trans-+/--2-aminocyclohexyl methanol hydrochloride, trans-+/--2-aminocyclohexyl methanol hydrochloride salt, cyclohexanemethanol, 2-amino-, hydrochloride, 1r,2r-rel
GCWPGEWXYDEQAY-NKWVEPMBSA-O
[(1R,2R)-2-aminocyclohexyl]methanol;hydrochloride
2724654
99+%
trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99+%
Videos
SDS
Documents

Documents

RUO – Research Use Only