missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hydroxycinnamic acid, 98+%

Catalog No. p-7081669
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1174014 25 g
AAA1174022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1174014 Supplier Thermo Scientific Chemicals Supplier No. A1174014
Only null left

CAS: 614-60-8 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 614-60-8
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00004379
InChI Key PMOWTIHVNWZYFI-AATRIKPKSA-N
Synonym 2-hydroxycinnamic acid, o-coumaric acid, trans-2-hydroxycinnamic acid, 2-coumaric acid, trans-o-hydroxycinnamic acid, trans-o-coumaric acid, 2-coumarate, e-o-hydroxycinnamic acid, 2-hydroxycinnamate, o-hydroxy-trans-cinnamic acid
PubChem CID 637540
ChEBI CHEBI:18125
IUPAC Name (E)-3-(2-hydroxyphenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC=C1O

Specifications

Melting Point ∼210°C (decomposition)
Quantity 25 g
Beilstein 1100900
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material trans-2-Hydroxycinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-386-4
RTECSNumber GD9090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only