missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenyl Valerate 94.0+%, TCI America™

Catalog No. H144425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H144425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H144425ML Supplier TCI America Supplier No. H144425ML
Only null left

Chemical Identifiers

CAS 56922-74-8
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
MDL Number MFCD00036547
InChI Key WDXAMNXWZLXISB-BQYQJAHWSA-N
Synonym Valeric Acid trans-2-Hexenyl Ester
PubChem CID 5352974
IUPAC Name (2E)-hex-2-en-1-yl pentanoate
SMILES CCCCC(=O)OC\C=C\CCC

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 184.28
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Hexenyl Valerate
EINECSNumber (2)-0760&(2)-0762
RTECSNumber SA3697500
TSCA Yes