accessibility menu, dialog, popup

UserName

H3COOCH3(E)

CAS RN 56922-74-8

IUPAC Name: (2E)-hex-2-en-1-yl pentanoate
Synonyms: trans-2-hexenyl valerate
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: WDXAMNXWZLXISB-BQYQJAHWSA-N
SMILES: CCCCC(=O)OC\C=C\CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results