Learn More
trans-2-Hexenoic acid, 96%
CAS: 13419-69-7 | C6H9O2 | 113.14 g/mol
$61.81 - $152.16
Chemical Identifiers
| CAS | 13419-69-7 |
|---|---|
| Molecular Formula | C6H9O2 |
| Molecular Weight (g/mol) | 113.14 |
| MDL Number | MFCD00002705 |
| InChI Key | NIONDZDPPYHYKY-SNAWJCMRSA-M |
| Synonym | trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e |
| PubChem CID | 5282707 |
| ChEBI | CHEBI:87721 |
| IUPAC Name | (E)-hex-2-enoic acid |
| SMILES | CCC\C=C\C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1357109
|
N/A | 10 g |
Each for $61.81
|
|
|||||
|
AAA1357118
|
N/A | 50 g |
Each for $152.16
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13419-69-7 | |
| 113.14 | |
| NIONDZDPPYHYKY-SNAWJCMRSA-M | |
| 5282707 | |
| (E)-hex-2-enoic acid |
| C6H9O2 | |
| MFCD00002705 | |
| trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e | |
| CHEBI:87721 | |
| CCC\C=C\C([O-])=O |
Specifications
| 13419-69-7 | |
| 0.965 | |
| 125°C (257°F) | |
| 1.4385 | |
| UN3261 | |
| trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e | |
| CCC\C=C\C([O-])=O | |
| 113.14 | |
| CHEBI:87721 | |
| 96% |
| 30°C to 35°C | |
| 217°C | |
| C6H9O2 | |
| MFCD00002705 | |
| 1720443 | |
| NIONDZDPPYHYKY-SNAWJCMRSA-M | |
| (E)-hex-2-enoic acid | |
| 5282707 | |
| 114.14 | |
| trans-2-Hexenoic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 236-528-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only