missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenoic acid, 96%

Catalog No. AAA1357118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1357118 50 g
AAA1357109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1357118 Supplier Thermo Scientific Chemicals Supplier No. A1357118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13419-69-7 | C6H9O2 | 113.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13419-69-7
Molecular Formula C6H9O2
Molecular Weight (g/mol) 113.14
MDL Number MFCD00002705
InChI Key NIONDZDPPYHYKY-SNAWJCMRSA-M
Synonym trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e
PubChem CID 5282707
ChEBI CHEBI:87721
IUPAC Name (E)-hex-2-enoic acid
SMILES CCC\C=C\C([O-])=O

Specifications

Melting Point 30°C to 35°C
Density 0.965
Boiling Point 217°C
Flash Point 125°C (257°F)
Refractive Index 1.4385
Quantity 50 g
UN Number UN3261
Beilstein 1720443
Formula Weight 114.14
Percent Purity 96%
Chemical Name or Material trans-2-Hexenoic acid
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 236-528-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only