missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenoic acid, 96%

CAS: 13419-69-7 | C6H9O2 | 113.14 g/mol

Supplier:  Thermo Scientific Chemicals A1357109

 View more versions of this product

Catalog No. AAA1357109


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-2-Hexenoic acid
30°C to 35°C
217°C
C6H9O2
MFCD00002705
UN3261
trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e
CCC\C=C\C([O-])=O
113.14
CHEBI:87721
96%
13419-69-7
0.965
125°C (257°F)
1.4385
10 g
1720443
NIONDZDPPYHYKY-SNAWJCMRSA-M
(E)-hex-2-enoic acid
5282707
114.14

Chemical Identifiers

13419-69-7
113.14
NIONDZDPPYHYKY-SNAWJCMRSA-M
5282707
(E)-hex-2-enoic acid
C6H9O2
MFCD00002705
trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e
CHEBI:87721
CCC\C=C\C([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 236-528-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only