missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenal 97.0+%, TCI America™

Catalog No. H034525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H034525ML 25 mL
H0345100ML 100 mL
H0345500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H034525ML Supplier TCI America Supplier No. H034525ML

Chemical Identifiers

CAS 6728-26-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00007008
InChI Key MBDOYVRWFFCFHM-SNAWJCMRSA-N
Synonym trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e
PubChem CID 5281168
ChEBI CHEBI:28913
IUPAC Name (E)-hex-2-enal
SMILES CCCC=CC=O

Specifications

Color Yellow
Boiling Point 146°C
Quantity 25 mL
UN Number 1988
Formula Weight 98.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Hexenal
EINECSNumber (2)-0520
RTECSNumber MP5900000
TSCA Yes
Recommended Storage Refrigerator