missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenal, 96%

CAS: 6728-26-3 | C6H10O | 98.145 g/mol

$71.49 - $213.60

Chemical Identifiers

CAS 6728-26-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00007008
InChI Key MBDOYVRWFFCFHM-SNAWJCMRSA-N
Synonym trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e
PubChem CID 5281168
ChEBI CHEBI:28913
IUPAC Name (E)-hex-2-enal
SMILES CCCC=CC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1151314
View Documents
Thermo Scientific Chemicals
A1151314
25 g
Each for $71.49
Only null left
 
AAA1151322
View Documents
Thermo Scientific Chemicals
A1151322
100 g
Each for $213.60
Only null left
 
Description

Description

trans-2-Hexenal is used in evaluation of quality of protected designation of origin virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. It was used in the multicomponent method for the determination of low-molecular-weight carbonyl compounds in biological samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-2-Hexenal is used in evaluation of quality of protected designation of origin virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. It was used in the multicomponent method for the determination of low-molecular-weight carbonyl compounds in biological samples.

Solubility
Not miscible in water.

Notes
Store at 4°C. Incompatible materials are bases, air, oxidizing agents, reducing agents and heat.
Specifications

Chemical Identifiers

6728-26-3
98.145
MBDOYVRWFFCFHM-SNAWJCMRSA-N
5281168
(E)-hex-2-enal
C6H10O
MFCD00007008
trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e
CHEBI:28913
CCCC=CC=O

Specifications

6728-26-3
37°C to 38°C (8 mmHg)
Sweet
1.447
25 g
1699684
trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e
MBDOYVRWFFCFHM-SNAWJCMRSA-N
(E)-hex-2-enal
5281168
98.15
trans-2-Hexenal
0.848
38°C (100°F)
C6H10O
MFCD00007008
UN1988
Air sensitive
Not miscible in water.
CCCC=CC=O
98.145
CHEBI:28913
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H226-H302-H332-H315-H319-H335
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P333+P313-P363-P370+P378q-P501c

H226-H302-H311-H317-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, FLAMMABLE, TOXIC, N.O.S.

EINECSNumber : 229-778-1

RTECSNumber : MP5900000

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only