missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexadecenoyl-L-carnitine, ≥95.0% (LC/MS), MilliporeSigma™ Supelco™

Catalog No. 111013768
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013768 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013768 Supplier Merck Emd Millipore Supplier No. 4100310MG

Specifications

CAS 26145-55-1
Linear Formula C23H43 NO4
Synonym (2 R)-3-Carboxy-N,N,N-trimethyl-2-[[(2 E)-1-oxo-2-hexadecen-1-yl]oxy]-1-propanaminium inner salt
Type Analytical Standard
Quantity 10 mg
Formula Weight 397.59
Percent Purity ≥95.0% (LC/MS)
Grade Analytical Standard