missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Heptenal 95.0+%, TCI America™

$139.81 - $376.54

Chemical Identifiers

CAS 18829-55-5
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00007010
InChI Key NDFKTBCGKNOHPJ-AATRIKPKSA-N
Synonym e-hept-2-enal, trans-2-heptenal, 2-heptenal, butylacrolein, 3-butylacrolein, 2-heptenal, e, e-2-hepten-1-al, 2-heptenal, 2e, beta-butylacrolein, trans-2-hepten-1-al
PubChem CID 5283316
IUPAC Name (E)-hept-2-enal
SMILES CCCCC=CC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H05795ML
View Documents
TCI America
H05795ML
5 mL
Each for $139.81
Only null left
 
H057925ML
View Documents
TCI America
H057925ML
25 mL
Each for $376.54
Only null left
 
Description

Description

Specifications

Chemical Identifiers

18829-55-5
112.172
NDFKTBCGKNOHPJ-AATRIKPKSA-N
5283316
CCCCC=CC=O
C7H12O
MFCD00007010
e-hept-2-enal, trans-2-heptenal, 2-heptenal, butylacrolein, 3-butylacrolein, 2-heptenal, e, e-2-hepten-1-al, 2-heptenal, 2e, beta-butylacrolein, trans-2-hepten-1-al
(E)-hept-2-enal

Specifications

18829-55-5
91°C
MFCD00007010
1988
NDFKTBCGKNOHPJ-AATRIKPKSA-N
(E)-hept-2-enal
5283316
≥95.0% (GC)
trans-2-Heptenal
Yellow
C7H12O
5 mL
e-hept-2-enal, trans-2-heptenal, 2-heptenal, butylacrolein, 3-butylacrolein, 2-heptenal, e, e-2-hepten-1-al, 2-heptenal, 2e, beta-butylacrolein, trans-2-hepten-1-al
CCCCC=CC=O
112.172
112.17
Liquid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : MJ8795000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'