missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Heptenal 95.0+%, TCI America™
Supplier: TCI America H05795ML
Specifications
| trans-2-Heptenal | |
| Yellow | |
| C7H12O | |
| 5 mL | |
| e-hept-2-enal, trans-2-heptenal, 2-heptenal, butylacrolein, 3-butylacrolein, 2-heptenal, e, e-2-hepten-1-al, 2-heptenal, 2e, beta-butylacrolein, trans-2-hepten-1-al | |
| CCCCC=CC=O | |
| 112.172 | |
| 112.17 | |
| Liquid |
| 18829-55-5 | |
| 91°C | |
| MFCD00007010 | |
| 1988 | |
| NDFKTBCGKNOHPJ-AATRIKPKSA-N | |
| (E)-hept-2-enal | |
| 5283316 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 18829-55-5 | |
| 112.172 | |
| NDFKTBCGKNOHPJ-AATRIKPKSA-N | |
| 5283316 | |
| CCCCC=CC=O |
| C7H12O | |
| MFCD00007010 | |
| e-hept-2-enal, trans-2-heptenal, 2-heptenal, butylacrolein, 3-butylacrolein, 2-heptenal, e, e-2-hepten-1-al, 2-heptenal, 2e, beta-butylacrolein, trans-2-hepten-1-al | |
| (E)-hept-2-enal |
Safety and Handling
RTECSNumber : MJ8795000
TSCA : Yes