missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Ethoxyethenyl-1-boronic acid pinacol ester, 95%

Catalog No. AAH3336703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3336703 1 g
AAH3336706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3336703 Supplier Thermo Scientific Chemicals Supplier No. H3336703

CAS: 1201905-61-4 | C10H19BO3 | 198.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1201905-61-4
Molecular Formula C10H19BO3
Molecular Weight (g/mol) 198.07
MDL Number MFCD09702441,MFCD09998813
InChI Key MRAYNLYCQPAZJN-UHFFFAOYSA-N
Synonym e-2-2-ethoxyvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-2-ethoxyvinylboronic acid pinacol ester, 2-e-2-ethoxyethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-ethoxyvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-1-ethoxyethene-2-boronic acid pinacol ester, 2-2-ethoxyethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-1-ethoxyethene-2-ylboronic acid pinacol ester, z-2-ethoxyethenyl boronic acid, pinacol ester, trans-2-ethoxyethenyl-1-boronic acid pinacol ester
PubChem CID 21973908
IUPAC Name 2-[(E)-2-ethoxyethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CCOC=CB1OC(C)(C)C(C)(C)O1

Specifications

Boiling Point 50°C (0.3 mmHg)
Refractive Index 1.446
Quantity 1 g
Formula Weight 198.07
Percent Purity 98%
Chemical Name or Material trans-2-Ethoxyethenyl-1-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only