accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1201905-61-4

BOOH3CCH3H3CH3COCH3(E/Z)

CAS RN 1201905-61-4

IUPAC Name: 2-(2-ethoxyethenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2-ethoxyethenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 198.07
Molecular Formula: C10H19BO3
InChi Key: MRAYNLYCQPAZJN-UHFFFAOYSA-N
SMILES: CCOC=CB1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.