missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Decenoic Acid 95.0+%, TCI America™

Catalog No. D00985ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00985ML 5 mL
D009825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00985ML Supplier TCI America Supplier No. D00985ML
Only null left

Chemical Identifiers

CAS 334-49-6
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00039530
InChI Key WXBXVVIUZANZAU-CMDGGOBGSA-N
Synonym 2-decenoic acid, trans-2-decenoic acid, e-2-decenoic acid, e-dec-2-enoic acid, unii-332t8th7b1, decenoic acid, 5 6-decenoic acid, trans-2-decylenic acid, trans-dec-2-enoic acid
PubChem CID 5282724
ChEBI CHEBI:50467
IUPAC Name (E)-dec-2-enoic acid
SMILES CCCCCCCC=CC(=O)O

Specifications

Color Yellow
Boiling Point 121°C
Quantity 5 mL
Formula Weight 170.25
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material trans-2-Decenoic Acid
EINECSNumber (2)-0609
TSCA No