CAS RN 334-49-6
CAS RN 334-49-6
IUPAC Name:
(2E)-dec-2-enoic acid
Synonyms:
trans-2-decenoic acid 2-decylenic acid (E)-2-decensaeure
Molecular Weight (g/mol):
170.25
Molecular Formula:
C10H18O2
InChi Key:
WXBXVVIUZANZAU-CMDGGOBGSA-N
SMILES:
CCCCCCC\C=C\C(O)=O
1
–
1
of
1
results
1
trans-2-Decenoic Acid 95.0+%, TCI America™
CAS: 334-49-6 Molecular Formula: C10H18O2 Molecular Weight (g/mol): 170.252 MDL Number: MFCD00039530 InChI Key: WXBXVVIUZANZAU-CMDGGOBGSA-N Synonym: 2-decenoic acid,trans-2-decenoic acid,e-2-decenoic acid,e-dec-2-enoic acid,unii-332t8th7b1,decenoic acid,5 6-decenoic acid,trans-2-decylenic acid,trans-dec-2-enoic acid PubChem CID: 5282724 ChEBI: CHEBI:50467 IUPAC Name: (E)-dec-2-enoic acid SMILES: CCCCCCCC=CC(=O)O