missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Decenal 93.0+%, TCI America™

Catalog No. p-7126844
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
D16425ML 5 mL
D164225ML 25 mL
2 options

Catalog No. D16425ML

Supplier: TCI America D16425ML

Only null left

Chemical Identifiers

CAS 3913-81-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00014679
InChI Key MMFCJPPRCYDLLZ-CMDGGOBGSA-N
Synonym trans-2-decenal, e-dec-2-enal, 2-decenal, decenal, e-2-decenal, 3-heptylacrolein, decenaldehyde, decylenic aldehyde, 2-decenal, 2e, 2-decenal, e
PubChem CID 5283345
IUPAC Name (2E)-dec-2-enal
SMILES CCCCCCC\C=C\C=O

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 mL
Formula Weight 154.25
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Decenal