missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Decenal, 95%

Catalog No. p-7044326
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0590106 5 g
AAL0590114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0590106 Supplier Thermo Scientific Chemicals Supplier No. L0590106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3913-81-3 | C10H18O | 154.25 g/mol

trans-2-Decenal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-2-Decenal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3913-81-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00014679
InChI Key MMFCJPPRCYDLLZ-CMDGGOBGSA-N
Synonym trans-2-decenal, e-dec-2-enal, 2-decenal, decenal, e-2-decenal, 3-heptylacrolein, decenaldehyde, decylenic aldehyde, 2-decenal, 2e, 2-decenal, e
PubChem CID 5283345
SMILES CCCCCCC\C=C\C=O

Specifications

Density 0.84
Boiling Point 117°C (17 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.4565
Quantity 5 g
Beilstein 1704445
Solubility Information Not miscible in water.
IUPAC Name (E)-dec-2-enal
Formula Weight 154.25
Percent Purity 95%
Chemical Name or Material trans-2-Decenal
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 223-474-2
RTECSNumber HE2070000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only