missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Boc-amino)cyclopropanemethanol, 97%

CAS: 170299-53-3 | C9H17NO3 | 187.24 g/mol

$365.42 - $1,032.70

Chemical Identifiers

CAS 170299-53-3
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD03844612
InChI Key OCKKMJSVLVALMF-UHFFFAOYNA-N
Synonym tert-butyl trans-2-hydroxymethyl cyclopropylcarbamate, trans-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl 1r,2r-2-hydroxymethyl cyclopropyl carbamate, tert-butyl trans-2-hydroxymethyl-cyclopropylcarbamate, 1r-2alpha-tert-butoxycarbonylamino cyclopropane-1beta-methanol, trans-2-hydroxymethyl-cyclopropyl-carbamic acid tert-butyl ester, tert-butyl n-1r,2r-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, trans
PubChem CID 24720934
IUPAC Name tert-butyl N-[(1R,2R)-2-(hydroxymethyl)cyclopropyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CC1CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62222MD
View Documents
Thermo Scientific Chemicals
H62222MD
250 mg N/A
Only null left
 
AAH6222203
View Documents
Thermo Scientific Chemicals
H6222203
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

170299-53-3
187.24
OCKKMJSVLVALMF-UHFFFAOYNA-N
24720934
CC(C)(C)OC(=O)NC1CC1CO
C9H17NO3
MFCD03844612
tert-butyl trans-2-hydroxymethyl cyclopropylcarbamate, trans-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl 1r,2r-2-hydroxymethyl cyclopropyl carbamate, tert-butyl trans-2-hydroxymethyl-cyclopropylcarbamate, 1r-2alpha-tert-butoxycarbonylamino cyclopropane-1beta-methanol, trans-2-hydroxymethyl-cyclopropyl-carbamic acid tert-butyl ester, tert-butyl n-1r,2r-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, trans
tert-butyl N-[(1R,2R)-2-(hydroxymethyl)cyclopropyl]carbamate

Specifications

170299-53-3
MFCD03844612
OCKKMJSVLVALMF-UHFFFAOYNA-N
tert-butyl N-[(1R,2R)-2-(hydroxymethyl)cyclopropyl]carbamate
187.24
187.23
trans-2-(Boc-amino)cyclopropanemethanol
C9H17NO3
tert-butyl trans-2-hydroxymethyl cyclopropylcarbamate, trans-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl 1r,2r-2-hydroxymethyl cyclopropyl carbamate, tert-butyl trans-2-hydroxymethyl-cyclopropylcarbamate, 1r-2alpha-tert-butoxycarbonylamino cyclopropane-1beta-methanol, trans-2-hydroxymethyl-cyclopropyl-carbamic acid tert-butyl ester, tert-butyl n-1r,2r-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, trans
CC(C)(C)OC(=O)NC1CC1CO
250 mg
24720934
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only