missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Boc-amino)cyclopropanemethanol, 97%

CAS: 170299-53-3 | C9H17NO3 | 187.24 g/mol

Supplier:  Thermo Scientific Chemicals H6222203

 View more versions of this product

Catalog No. AAH6222203

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

trans-2-(Boc-amino)cyclopropanemethanol
C9H17NO3
tert-butyl trans-2-hydroxymethyl cyclopropylcarbamate, trans-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl 1r,2r-2-hydroxymethyl cyclopropyl carbamate, tert-butyl trans-2-hydroxymethyl-cyclopropylcarbamate, 1r-2alpha-tert-butoxycarbonylamino cyclopropane-1beta-methanol, trans-2-hydroxymethyl-cyclopropyl-carbamic acid tert-butyl ester, tert-butyl n-1r,2r-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, trans
CC(C)(C)OC(=O)NC1CC1CO
1 g
24720934
97%
170299-53-3
MFCD03844612
OCKKMJSVLVALMF-UHFFFAOYNA-N
tert-butyl N-[(1R,2R)-2-(hydroxymethyl)cyclopropyl]carbamate
187.24
187.23

Chemical Identifiers

170299-53-3
187.24
OCKKMJSVLVALMF-UHFFFAOYNA-N
24720934
CC(C)(C)OC(=O)NC1CC1CO
C9H17NO3
MFCD03844612
tert-butyl trans-2-hydroxymethyl cyclopropylcarbamate, trans-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl 1r,2r-2-hydroxymethyl cyclopropyl carbamate, tert-butyl trans-2-hydroxymethyl-cyclopropylcarbamate, 1r-2alpha-tert-butoxycarbonylamino cyclopropane-1beta-methanol, trans-2-hydroxymethyl-cyclopropyl-carbamic acid tert-butyl ester, tert-butyl n-1r,2r-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, trans
tert-butyl N-[(1R,2R)-2-(hydroxymethyl)cyclopropyl]carbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only