missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%

CAS: 1212272-03-1 | C13H15NO4 | 249.27 g/mol

Supplier:  Thermo Scientific Chemicals H6291903

Quantity 1 g

 View more versions of this product

Catalog No. AAH6291903


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1212272-03-1
249.27
ZNRUEEQIUVKKBL-UHFFFAOYNA-N
24720935
OC(=O)C1CCC1NC(=O)OCC1=CC=CC=C1
C13H15NO4
MFCD03844619
trans-2-benzyloxycarbonylaminocyclobutanecarboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, trans-2-cbz-amino cyclobutane-1-carboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane carboxylic acid, 1r,2r-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid
(1R,2R)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid

Specifications

trans-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid
C13H15NO4
trans-2-benzyloxycarbonylaminocyclobutanecarboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, trans-2-cbz-amino cyclobutane-1-carboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane carboxylic acid, 1r,2r-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid
OC(=O)C1CCC1NC(=O)OCC1=CC=CC=C1
1 g
24720935
97%
1212272-03-1
MFCD03844619
ZNRUEEQIUVKKBL-UHFFFAOYNA-N
(1R,2R)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid
249.27
249.26
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions