missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%

Catalog No. p-7041641
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62919MD 250 mg
AAH6291903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62919MD Supplier Thermo Scientific Chemicals Supplier No. H62919MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1212272-03-1 | C13H15NO4 | 249.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1212272-03-1
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.27
MDL Number MFCD03844619
InChI Key ZNRUEEQIUVKKBL-UHFFFAOYNA-N
Synonym trans-2-benzyloxycarbonylaminocyclobutanecarboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, trans-2-cbz-amino cyclobutane-1-carboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane carboxylic acid, 1r,2r-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid
PubChem CID 24720935
IUPAC Name (1R,2R)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid
SMILES OC(=O)C1CCC1NC(=O)OCC1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 249.26
Percent Purity 97%
Chemical Name or Material trans-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only