missing translation for 'onlineSavingsMsg'
Learn More

trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline 95.0+%, TCI America™

Catalog No. D4652200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4652200MG 200 mg
D46521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4652200MG Supplier TCI America Supplier No. D4652200MG
Only null left

Chemical Identifiers

CAS 908294-68-8
Molecular Formula C18H22N2
Molecular Weight (g/mol) 266.39
MDL Number MFCD06669887
InChI Key BABQFMPURPKJNW-UHFFFAOYSA-N
PubChem CID 1284716
IUPAC Name N-[2,6-bis(propan-2-yl)phenyl]-1-(pyridin-2-yl)methanimine
SMILES CC(C)C1=CC=CC(C(C)C)=C1N=CC1=CC=CC=N1

Specifications

Quantity 200 mg
Formula Weight 266.39
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline