missing translation for 'onlineSavingsMsg'
Learn More

trans-2,5-Dimethylpiperazine 99.0+%, TCI America™

Catalog No. D07635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D07635G 5 g
D076325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D07635G Supplier TCI America Supplier No. D07635G

Chemical Identifiers

CAS 2815-34-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00070516
InChI Key NSMWYRLQHIXVAP-OLQVQODUSA-N
Synonym trans-2,5-dimethylpiperazine, 2r,5s-2,5-dimethylpiperazine, 2s,5r-2,5-dimethylpiperazine, 2s,5r-2,5-dimethyl-piperazine, piperazine, 2,5-dimethyl-, 2r,5s-rel, 2r,5s-2,5-dimethylpiperazin, piperazine, 2,5-dimethyl-, trans, 2r,5s-2,5-dimethyl-piperazine, pubchem15878, 2,5-dimethylpiperazine #
PubChem CID 220672
IUPAC Name (2R,5S)-2,5-dimethylpiperazine
SMILES CC1CNC(CN1)C

Specifications

Melting Point 116°C
Color White-Yellow
Boiling Point 162°C
Quantity 5 g
UN Number 2926
Formula Weight 114.19
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-2,5-Dimethylpiperazine
EINECSNumber (5)-0955
TSCA No