accessibility menu, dialog, popup

UserName

CH3(S)NH2(R)CH3H2N

CAS RN 2815-34-1

IUPAC Name: (2R,5S)-2,5-dimethylpiperazine-1,4-diium
Synonyms: (2R,5S)-2,5-dimethylpiperazine-1,4-diium
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: NSMWYRLQHIXVAP-OLQVQODUSA-P
SMILES: C[C@H]1C[NH2+][C@H](C)C[NH2+]1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)