missing translation for 'onlineSavingsMsg'
Learn More

trans-2,4-Dichlorocinnamic Acid 98.0+%, TCI America™

Catalog No. D25885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25885G 5 g
D258825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25885G Supplier TCI America Supplier No. D25885G
Only null left

Chemical Identifiers

CAS 20595-45-3
Molecular Formula C9H6Cl2O2
Molecular Weight (g/mol) 217.045
MDL Number MFCD00004373
InChI Key MEBWABJHRAYGFW-DUXPYHPUSA-N
Synonym 2,4-dichlorocinnamic acid, trans-2,4-dichlorocinnamic acid, e-3-2,4-dichlorophenyl acrylic acid, cinnamic acid, 2,4-dichloro, 3-2,4-dichlorophenyl-2-propenoic acid, unii-c09kk5d2t2, 2-propenoic acid, 3-2,4-dichlorophenyl-, 2e, 2e-3-2,4-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,4-dichlorophenyl, 2e-3-2,4-dichlorophenyl prop-2-enoic acid
PubChem CID 688026
IUPAC Name (E)-3-(2,4-dichlorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)C=CC(=O)O

Specifications

Melting Point 236°C
Color White
Quantity 5 g
Formula Weight 217.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-2,4-Dichlorocinnamic Acid
RTECSNumber GD8575000
TSCA No