CAS RN 20595-45-3
IUPAC Name:
(2E)-3-(2,4-dichlorophenyl)prop-2-enoic acid
Synonyms:
2,4-dichlorocinnamic acid
Molecular Weight (g/mol):
217.05
Molecular Formula:
C9H6Cl2O2
InChi Key:
MEBWABJHRAYGFW-DUXPYHPUSA-N
SMILES:
OC(=O)\C=C\C1=CC=C(Cl)C=C1Cl
1
–
1
of
1
results
1
trans-2,4-Dichlorocinnamic Acid 98.0+%, TCI America™
CAS: 20595-45-3 Molecular Formula: C9H6Cl2O2 Molecular Weight (g/mol): 217.045 MDL Number: MFCD00004373 InChI Key: MEBWABJHRAYGFW-DUXPYHPUSA-N Synonym: 2,4-dichlorocinnamic acid,trans-2,4-dichlorocinnamic acid,e-3-2,4-dichlorophenyl acrylic acid,cinnamic acid, 2,4-dichloro,3-2,4-dichlorophenyl-2-propenoic acid,unii-c09kk5d2t2,2-propenoic acid, 3-2,4-dichlorophenyl-, 2e,2e-3-2,4-dichlorophenyl acrylic acid,2-propenoic acid, 3-2,4-dichlorophenyl,2e-3-2,4-dichlorophenyl prop-2-enoic acid PubChem CID: 688026 IUPAC Name: (E)-3-(2,4-dichlorophenyl)prop-2-enoic acid SMILES: C1=CC(=C(C=C1Cl)Cl)C=CC(=O)O