missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(4-chlorophenyl)vinylboronic acid, 97%

Catalog No. p-4228545
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431340010 1 g Glass bottle
AC431340100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC431340010 Supplier Thermo Scientific Chemicals Supplier No. 431340010

CAS: 154230-29-2 | C8H8BClO2 | 182.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154230-29-2
Molecular Formula C8H8BClO2
Molecular Weight (g/mol) 182.41
MDL Number MFCD02093767
InChI Key HWSDRAPTZRYXHN-AATRIKPKSA-N
Synonym trans-2-4-chlorophenyl vinylboronic acid, 4-chlorostyryl boronic acid, e-2-4-chlorophenyl ethenyl boronic acid, e-4-chlorostyrylboronic acid, boronic acid, 1e-2-4-chlorophenyl ethenyl, e-2-4-chlorophenyl ethenylboronic acid, trans-2-4-chlorophenyl ethenylboronic acid, rarechem al ba 0169, e-2-4-chlorophenyl vinyl boronic acid, 4-chlorostyrylboronic acid
PubChem CID 642694
IUPAC Name [(E)-2-(4-chlorophenyl)ethenyl]boronic acid
SMILES OB(O)\C=C\C1=CC=C(Cl)C=C1

Specifications

Melting Point 133.0°C to 134.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 182.41
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material trans-2-(4-chlorophenyl)vinylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only