missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene, 97%

Catalog No. AC432030010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432030010 1 g Glass bottle
AC432030050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432030010 Supplier Thermo Scientific Chemicals Supplier No. 432030010

CAS: 83947-56-2 | C14H19BO2 | 230.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83947-56-2
Molecular Formula C14H19BO2
Molecular Weight (g/mol) 230.11
MDL Number MFCD03453666
InChI Key ARAINKADEARZLZ-ZHACJKMWSA-N
Synonym e-4,4,5,5-tetramethyl-2-styryl-1,3,2-dioxaborolane, e-phenylethenylboronic acid, pinacol ester, trans-styrylboronic acid, pinacol ester, beta-styrylboronic acid pinacol ester, trans-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl styrene, 4,4,5,5-tetramethyl-2-styryl-1,3,2-dioxaborolane, trans-beta-styrylboronic acid pinacol ester, trans-2-phenylvinylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-e-2-phenylethenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-2-phenylvinyl-1,3,2-dioxaborolane
PubChem CID 5708413
IUPAC Name 4,4,5,5-tetramethyl-2-[(E)-2-phenylethenyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)\C=C\C1=CC=CC=C1

Specifications

Color Colorless to Yellow
Boiling Point 89°C to 93°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 230.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only