missing translation for 'onlineSavingsMsg'
Learn More

trans-2,3-Dimethoxycinnamic Acid 98.0+%, TCI America™

Catalog No. d232625g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
D232625G 25 g
1 options

Catalog No. D232625G

Supplier: TCI America D232625G

Only null left

Chemical Identifiers

CAS 7345-82-6
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00004376
InChI Key QAXPUWGAGVERSJ-VOTSOKGWSA-N
Synonym 2,3-dimethoxycinnamic acid, trans-2,3-dimethoxycinnamic acid, 2e-3-2,3-dimethoxyphenyl acrylic acid, e-3-2,3-dimethoxyphenyl acrylic acid, 2,3-dimethoxy cinnamic acid, 3-2,3-dimethoxyphenyl acrylic acid, 2e-3-2,3-dimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,3-dimethoxyphenyl, e-3-2,3-dimethoxyphenyl prop-2-enoic acid, e-3-2,3-dimethoxy-phenyl-acrylic acid
PubChem CID 735842
IUPAC Name (E)-3-(2,3-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=CC(=C1OC)C=CC(=O)O

Specifications

Melting Point 180°C
Color White
Quantity 25 g
Formula Weight 208.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans-2,3-Dimethoxycinnamic Acid