accessibility menu, dialog, popup

UserName

CAS RN 7345-82-6

IUPAC Name: 3-(2,3-dimethoxyphenyl)prop-2-enoic acid
Synonyms: 3-(2,3-dimethoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: QAXPUWGAGVERSJ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C=CC(O)=O)=C1OC
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)