missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 194°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Synonym | trans-4′-Carboxycotinine |
| Formula Weight | 220.23 |
| Percent Purity | ≥95.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone |
Safety and Handling
| TSCA | No |